Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe | 59880-97-6 | 99.90% | 437.55 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP17445MG
Encompass_Preferred
N-Formyl-Met-Leu-Phe (fMLP) is a chemotactic peptide and specific ligand for the N-formyl peptide receptor (FPR). It inhibits TNF-alpha secretion and promotes osteoblastic commitment. Binding to FPR triggers various biochemical events, leading to cellular activation.
  • Specific ligand for N-formyl peptide receptor (FPR)
  • Inhibits TNF-alpha secretion
  • Promotes osteoblastic and suppresses adipogenic commitment
  • Stimulates osteogenesis and mineralization
  • Induces proinflammatory cytokine gene expression
  • Promotes bone formation in zebrafish and rabbits
Catalog No. 50-004-24450