Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe (fMLP) | 59880-97-6 | 99.9% | 437.55 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP022450MG
Encompass_Preferred
N-Formyl-Met-Leu-Phe (fMLP) is a chemotactic peptide that acts as a specific ligand for the N-formyl peptide receptor (FPR). It is also reported to inhibit TNF-alpha secretion.
  • Specific ligand for N-formyl peptide receptor (FPR).
  • Inhibits TNF-alpha secretion.
  • Promotes osteoblastic commitment and suppresses adipogenic commitment; stimulates osteogenesis and mineralization.
  • Induces proinflammatory cytokine gene expression in human peripheral blood monocytes; synergizes with bacterial products like LPS.
  • Promotes bone formation in zebrafish and rabbits, increasing Runx2 expression.
  • Induces release of calprotectin from polymorphonuclear cells (PMN) in vitro.
Catalog No. 50-004-25364