Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe-Lys | 67247-11-4 | 99.8% | 565.73 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP174410MG
Encompass_Preferred
N-Formyl-Met-Leu-Phe-Lys (fMLFK) is a peptide that acts as a potent and selective agonist of FPR1, with EC50s of 3.5 nM for FPR1, 6.7 μM for FPR2, and 0.88 μM for FPR2-D2817.32G.
  • Acts as a potent and selective agonist of FPR1
  • EC50 of 3.5 nM for FPR1
  • EC50 of 6.7 μM for FPR2
  • EC50 of 0.88 μM for FPR2-D2817.32G
  • Classified as a peptide
  • Relevant in inflammation and immunology research
Catalog No. 50-004-23704