Learn More
Medchemexpress LLC N-Formyl-MMYALF | 863418-59-1 | >99.78% | 803.00 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1073810MG
Encompass_Preferred
N-Formyl-MMYALF is a potent mitochondrial N-formyl peptide (mtFP) that depletes calcium ions in the endoplasmic reticulum. It can inhibit the FPR-1-mediated chemotactic response of polymorphonuclear leukocytes (PMNs) to bacterial peptides.
  • Potent mitochondrial N-formyl peptide
  • Depletes calcium ions in the endoplasmic reticulum
  • Inhibits FPR-1-mediated chemotactic response of polymorphonuclear leukocytes to bacterial peptides
Catalog No. 50-004-25687