Learn More
Medchemexpress LLC N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys | 71901-21-8 | 98.9% | 824.02 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1591
Encompass_Preferred
N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys is a formyl peptide receptor (FPR) agonist. It serves as a laboratory chemical for the manufacture of substances, particularly in research concerning inflammation or immune system diseases. This product is intended for research use only by experienced personnel.
  • Formyl peptide receptor (FPR) agonist
  • Peptide and peptide derivative
  • Functions as an inhibitor or substrate
  • Applicable to inflammation and immune system disease research
  • Appears as a solid, white to off-white powder
  • Recommended for sealed storage away from moisture
  • Stable for 2 years as powder at -80°C and 1 year at -20°C
  • Stable for 6 months in solvent at -80°C and 1 month at -20°C
Catalog No. 50-004-26955