Learn More
Medchemexpress LLC N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys | 71901-21-8 | 98.9% | 824.02 | 50 MG

Supplier: Medchemexpress LLC HYP159150MG
N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys TFA (For-Nle-Leu-Phe-Nle-Tyr-Lys-OH TFA) is a formyl peptide receptor (FPR) agonist. It is suitable for laboratory research, appearing as a solid with a white to off-white color. The product is for research use only.
- Formyl peptide receptor (FPR) agonist
- Appearance: Solid, white to off-white
- Sequence: N-Formyl-{NLE}-Leu-Phe-{NLE}-Tyr-Lys
- Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
- Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.