Learn More
Medchemexpress LLC N-Formylcytisine | 53007-06-0 | 10 MG

Supplier: Medchemexpress LLC HYN762510MG
N-Formylcytisine is a cytisine-type alkaloid derived from the stem bark of Maackia amurensis. It is intended for research use only.
- CAS number: 53007-06-0
- Molecular Formula: C12H14N2O2
- Molecular Weight: 218.25
- Appearance: Solid, white to off-white
- Purity: 99.17%
- Solubility in DMSO: 100 mg/mL (requires ultrasonication)
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.