Learn More
Medchemexpress LLC N-Formylglycine | 2491-15-8 | 95.0% | 1 ML

Supplier: Medchemexpress LLC HY12873510MM/1ML
N-Formylglycine is an endogenous metabolite intended for research use only.
- Endogenous metabolite
- Molecular weight of 103.08
- Molecular formula: C3H5NO3
- Purity of 95.0%
- Appears as a white to off-white solid
- Recommended storage for powder: 4°C, sealed, away from moisture
- Recommended storage for in-solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.