Learn More
Medchemexpress LLC N-(furan-2-ylmethyl)-3-[4-[methyl(propyl)amino]-6-(trifluoromethyl)pyrimidin-2-yl]sulfanylpropanamide | 2313525-20-9 | 99.8% | C17H21F3N4O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13222210MG
Encompass_Preferred
SET2 is a research compound that functions as a selective TRPV2 antagonist used to investigate TRPV2-mediated signaling in biochemical and cell-based assays. It has documented inhibitory potency and has been used in studies of calcium signaling and cell migration.
  • Selective TRPV2 antagonist (IC50 = 0.46 μM)
  • Useful in biochemical and cell-based assays
  • Reduces lysophosphatidic acid-induced cytosolic calcium increase
  • Reported to suppress prostate cancer cell migration
  • High purity suitable for research applications
  • Molecular formula: C17H21F3N4O2S; molecular weight: 402.43 g/mol
  • Available in multiple small-scale pack sizes for laboratory use
Catalog No. 50-002-61059