Learn More
Medchemexpress LLC N-hexacosane-d54 | 1219803-91-4 | 99.5% | 421.04 g/mol | C26D54 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW131352S25MG
Encompass_Preferred
N-Hexacosane-d54 is a deuterium-labeled analogue of n-hexacosane used as an isotopic tracer and internal standard for analytical and research applications. It provides a defined mass shift for reliable quantitation in mass spectrometry and gas chromatography and is supplied for laboratory research use only.
  • Deuterium labeled (d54) for clear mass spectral distinction.
  • High purity 99.5% supports accurate quantitation.
  • Stable, nonpolar hydrocarbon compatible with GC and MS methods.
  • Suitable as a tracer and internal standard in analytical workflows.
  • Available in small packaged quantities for laboratory use.
Catalog No. 50-002-82996