Learn More
Medchemexpress LLC N-hydroxy-4-[(8-quinolinylamino)methyl]benzamide | 2151853-97-1 | 99.8% | 293.32 g·mol⁻¹ | C17H15N3O2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1239765MG
Encompass_Preferred
MPT0G211 is a potent, orally active, and selective histone deacetylase 6 (HDAC6) inhibitor (CAS 2151853-97-1) used for preclinical research. It exhibits sub-nanomolar potency (IC50 = 0.291 nM), high selectivity for HDAC6, and reported blood-brain barrier penetration. The compound is supplied as solid quantities and as a 10 mM solution in DMSO, and is accompanied by a datasheet, SDS, and certificate of analysis for research use.
  • Potent HDAC6 inhibition (IC50 = 0.291 nM).
  • High selectivity for HDAC6 (>1000-fold vs other isoforms).
  • Reported blood-brain barrier penetration.
  • Available as solid and 10 mM solution in DMSO; common pack sizes 5 mg-100 mg.
  • High reported purity (~99.8%).
  • Molecular formula C17H15N3O2; molecular weight 293.32 g·mol⁻¹.
  • Provided with datasheet, SDS, and certificate of analysis.
Catalog No. 50-002-37453