Learn More
Medchemexpress LLC N-hydroxy-7-(naphthalen-2-ylthio)heptanamide | 926908-04-5 | MFCD10568146 | 99.8% | 303.42 | C17H21NO2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11867250MG
Encompass_Preferred
HNHA (N-hydroxy-7-(naphthalen-2-ylthio)heptanamide) is a potent histone deacetylase (HDAC) inhibitor supplied for research use. It induces histone hyperacetylation, arrests the cell cycle at G1/S via p21 induction, and has shown inhibition of tumor growth and tumor neovascularization in preclinical studies.
  • Potent HDAC inhibitor (IC50 = 100 nM).
  • Induces histone hyperacetylation and p21-mediated G1/S arrest.
  • Shown to inhibit tumor growth and tumor neovascularization in preclinical models.
  • High purity (99.76%).
  • Molecular weight 303.42 Da; formula C17H21NO2S.
  • Supplied as a solid with recommended storage at -20°C (powder) and -80°C (in solvent) as applicable.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-29543