Learn More
Medchemexpress LLC N-(hydroxymethyl)acetamide | 625-51-4 | MFCD00014417 | 96.5% | 89.09 g·mol-1 | C3H7NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW024564
Encompass_Preferred
N-(Hydroxymethyl)acetamide is a small organic compound (C3H7NO2, MW 89.09 g·mol-1) used as a reagent and model molecule in biochemical and physical chemistry research. It is suitable for studies of hydrogen bonding, dimerization, mechanistic investigations, and small-scale synthetic applications.
  • High purity (96.5%).
  • Low molecular weight and simple structure for model studies.
  • Soluble in common organic solvents and water.
  • Useful for hydrogen-bonding and dimerization investigations.
  • Available in laboratory-scale quantities for research use.
Catalog No. 50-003-03550