Learn More
Medchemexpress LLC N-(hydroxymethyl)acetamide | 625-51-4 | MFCD00014417 | 96.5% | 89.09 g/mol | C3H7NO2 | 5 G

Supplier: Medchemexpress LLC HYW0245645G
N-(Hydroxymethyl)acetamide is a low-molecular-weight biochemical reagent used as a model molecule in studies of hydrogen bonding and molecular interactions. It is supplied as a characterized solid with available analytical documentation for laboratory and materials research applications.
- Small, well-characterized molecule suitable for hydrogen-bonding studies.
- Molecular formula C3H7NO2; molecular weight 89.09 g/mol.
- Purity 96.46% as reported on the certificate of analysis.
- Available in multiple pack sizes, including 5 g, for flexible lab use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.