Learn More
Medchemexpress LLC N-(hydroxymethyl)acetamide | 625-51-4 | MFCD00014417 | 96.5% | 89.09 g/mol | C3H7NO2 | 50 G

Supplier: Medchemexpress LLC HYW02456450G
N-(Hydroxymethyl)acetamide is a small-molecule biochemical reagent supplied as a solid for laboratory research. The compound has CAS 625-51-4, molecular formula C3H7NO2, and a molecular weight of 89.09 g/mol. The manufacturer lists a stated purity of 96.46%, making it appropriate for assay reagents and use as a synthetic intermediate in research settings.
- Small-molecule reagent for biochemical assays
- Provided as a solid
- Molecular formula C3H7NO2 and molecular weight 89.09 g/mol
- Stated purity of 96.46%
- Suitable for research and synthetic intermediate use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.