Learn More
Medchemexpress LLC N-Isobutyrylguanosine | 64350-24-9 | 99.7% | 353.33 | 100 G
SDP
Supplier:  Medchemexpress LLC HY43057100G
Encompass_Preferred
iBu-rG (N-Isobutyrylguanosine) is a nucleoside compound that can be used to synthesize oligoribonucleotides.
  • Solid appearance
  • White to off-white color
  • Shipping at room temperature in continental US
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-04410