Learn More
Medchemexpress LLC N-isovalerylglycine-d9 | 1330037-21-2 | MFCD09953741 | 99.0% | 168.24 g/mol | C7H4D9NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015464S1MG
Encompass_Preferred
N-Isovalerylglycine-d9 is the deuterium-labeled isotopologue of N-isovaleroylglycine provided as a stable-isotope reagent for analytical applications. It is used as an internal standard or tracer in LC-MS/MS and metabolomics workflows for accurate quantification of isovaleroylglycine and related metabolites, and is applicable to biomarker research and method validation in metabolic studies.
  • Deuterium-labeled internal standard for LC-MS/MS and metabolomics.
  • High chemical purity, suitable for quantitative analysis.
  • Stable isotopic tracer for method validation and recovery studies.
  • Soluble in DMSO for convenient stock preparation.
  • Available in small sizes tailored to analytical needs.
Catalog No. 50-003-02790