Learn More
Medchemexpress LLC N-Mal-N-bis(PEG2-NH-Boc) | 2128735-26-0 | C29H50N4O11 | 100 MG

Supplier: Medchemexpress LLC HY140574100MG
N-Mal-N-bis(PEG2-NH-Boc) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to degrade target proteins by harnessing the ubiquitin-proteasome system, connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker. This compound is intended for research use only.
- Molecular formula: C29H50N4O11
- Molecular weight: 630.73
- CAS number: 2128735-26-0
- Target: PROTAC Linkers
- Pathway: PROTAC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.