Learn More
Medchemexpress LLC N-Mal-N-bis(PEG4-NH-Boc) | 2128735-27-1 | 97.0% | C37H66N4O15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140575100MG
Encompass_Preferred
N-Mal-N-bis(PEG4-NH-Boc) is a PEG-based PROTAC linker, intended for research use only. This compound can be utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Suitable for PROTAC synthesis
  • Supports the study of PROTACs that selectively degrade target proteins
  • For research use only
  • Exhibits in vitro biological activity
Catalog No. 50-003-53058