Learn More
Medchemexpress LLC N-Mal-N-bis(PEG4-NH-Boc) | 2128735-27-1 | 97.0% | C37H66N4O15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14057550MG
Encompass_Preferred
N-Mal-N-bis(PEG4-NH-Boc) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-53182