Learn More
Medchemexpress LLC N-Me-L-Ala-maytansinol | 77668-69-0 | MFCD00864836 | 99.9% | 650.16 g/mol | C32H44ClN3O9 | 100 MG

Supplier: Medchemexpress LLC HY126663100MG
N-Me-L-Ala-maytansinol is a maytansinoid cytotoxin supplied for research use as a hydrophobic, cell-permeable payload for constructing antibody-drug conjugates (ADCs).
- Hydrophobic, cell-permeable maytansinoid payload for ADC development.
- CAS 77668-69-0; molecular formula C32H44ClN3O9; molecular weight 650.16 g/mol.
- High purity (99.9%) suitable for research and conjugation.
- Available in small-scale quantities, including 100 MG.
- Storage: powder at -20°C (3 years) or 4°C (2 years); store solutions at -80°C for long-term use.
- Solubility: soluble in DMSO (100 mg/mL) with ultrasonic aid; several in vivo vehicle formulations reported.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.