Learn More
AdipoGen N-Methyl-1-deoxynojirimycin
SDP
Supplier:  AdipoGen BVT0130M001
Encompass_Preferred
Chemical. CAS 69567-10-8. Formula C7H15NO4. MW 177.2. Isolated from Streptomyces sp. alpha-Glucosidase inhibitor. Inhibitor of glycoprotein processing. Strong glycosyltransferase GTF inhibitor. Angiogenesis inhibitor. Reduces the size of myocardial infarcts.
Catalog No. 50-256-2091