Learn More
Medchemexpress LLC N-methyl-2-(pyridin-2-yl)-N-(2-(pyridin-2-yl)ethyl)ethanamine | 5452-87-9 | MFCD28157290 | 98.4% | 241.33 g/mol | C15H19N3 | 50 MG

Supplier: Medchemexpress LLC HY10749550MG
Betahistine EP Impurity C is an analytical reference standard used for identification and quantification of a betahistine-related impurity in pharmaceutical analysis. It is supplied as a characterized chemical with defined purity and supporting documentation for laboratory research and analytical method development.
- Available in 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg packages.
- Purity 98.4% as reported.
- Molecular formula C15H19N3 and molecular weight 241.33 g/mol.
- Provided with data sheet, certificate of analysis, and safety data sheet.
- Intended for analytical and research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.