Learn More
Abcam N-Methyl-4-phenylpyridinium Iodide (MPP+), Dopaminergic selective neuroto x in, 250MG
Supplier:  Abcam AB144783250MG
Encompass_Preferred
MW 297.13 Da, Purity >98%. Dopaminergic selective neurotoxin. Active MPTP metabolite. Interferes with oxidative phosphorylation and induces mitochondrial dysfunction. Deplete cellular ATP. Shows Parkinson effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3273