Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methyl-β-carboline-3-carboxamide | 78538-74-6 | MFCD00055075 | 99.4% | 225.25 | C13H11N3O | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10099110MM 1ML

Encompass_Preferred

FG 7142 is a beta-carboline benzodiazepine inverse agonist used as a research reagent to study GABAA receptor function. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid sizes to support different experimental needs.

  • Benzodiazepine inverse agonist that modulates GABAA receptor activity.
  • Supplied as 10 mM solution in DMSO, 1 mL, and as solid quantities.
  • High reported purity (≈99.4%).
  • CAS number 78538-74-6 for unambiguous identification.
  • Molecular weight 225.25 g/mol.
  • Intended for research use; follow appropriate safety procedures.

Catalog No. 50-002-12105


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.