Learn More
Sigma Aldrich Fine Chemicals Biosciences N Methyl D aspartic acid 91G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M32621G
Encompass
N-Methyl-D-aspartic acid (NMDA) is an endogenously generated molecule in rat nervous system and endocrine glands. NMDA is present at low level (nmol/g) in the adenohypophysis hypothalamus brain and testis. NMDA is derived from D-Asp by an S-adenosylmethionine-dependent enzyme also referred to as NMDA synthase.
Catalog No. 50-291-4555