Learn More
Medchemexpress LLC N-methyl-DL-aspartic acid | 17833-53-3 | MFCD00065918 | 98.0% | 147.13 g/mol | C5H9NO4 | 1 ML

Supplier: Medchemexpress LLC HYW01750010MM/1ML
N-Methyl-DL-aspartic acid is a glutamate analogue and selective NMDA receptor agonist used in neuroscience and neurological disease research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid material for laboratory studies; intended for research use only.
- Ready-to-use 10 mM solution in DMSO (1 mL).
- Also available as solid in multiple pack sizes.
- Purity 98.0%.
- Molecular weight 147.13 g/mol.
- Molecular formula C5H9NO4.
- CAS number 17833-53-3.
- Useful for studying NMDA receptor-mediated signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.