Learn More
Medchemexpress LLC N-methyl-L-alanine | 3913-67-5 | MFCD00037241 | 98.0% | 103.12 g/mol | C4H9NO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01592610G
Encompass_Preferred
H-N-Me-Ala-OH is an N-methylated alanine derivative supplied as a white to off-white solid for research use in peptide synthesis and related chemical biology applications. It has molecular formula C4H9NO2, a molecular weight of 103.12 g/mol, and is typically provided at 98.0% purity.
Catalog No. 50-003-00563