Learn More
Cayman Chemical N-methyl LeukotrIen C4 100ug
SDP
Supplier:  Cayman Chemical 13390100 UG
Encompass_Preferred
A stable analog of LTC4 that does not readily metabolize to LTD4 and LTE4 at the expense of marginally reduced biological activity
Catalog No. 50-295-5960