Learn More
Medchemexpress LLC N-methylacetamide-d1 | 3669-70-3 | MFCD03428088 | 99.4% | 74.10 g/mol | C3H6DNO | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW020018S150MG
Encompass_Preferred
N-methylacetamide-d1 is the deuterium-labeled analogue of N-methylacetamide provided as an analytical reagent for use as an internal standard or tracer in spectroscopic and mass-spectrometric assays. Supplied as a high-purity isotopically labeled material, it is intended for laboratory research applications and quantitative analyses.
  • Deuterium-labeled (d1) isotopologue for accurate mass spectral quantification.
  • High purity (>99%) for reliable analytical performance.
  • Available in small milligram quantities suitable for tracer studies.
  • Stable under recommended storage conditions for extended shelf life.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS analyses.
Catalog No. 50-003-01468