Learn More
Medchemexpress LLC N-methylacetamide-d6 | 3669-73-6 | 98.6% | 79.13 | C3HD6NO | 100 MG

Supplier: Medchemexpress LLC HYW020018S100MG
N-methylacetamide-d6 is the deuterium-labeled isotopologue of N-methylacetamide supplied as a stable isotope reagent for analytical and research applications. It is provided in laboratory-scale quantities and is accompanied by documentation for quality verification.
- Deuterium-labeled isotopologue (d6) for NMR and mass spectrometry.
- High isotopic and chemical purity suitable for analytical standards.
- Purity: 98.6%.
- Molecular formula C3HD6NO; molecular weight 79.13.
- Available in multiple small package sizes for research use.
- Datasheet, certificate of analysis, and SDS available for verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.