Learn More
Medchemexpress LLC N-Methylbenzylamine | 103-67-3 | 121.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW007426S
Encompass_Preferred
N-methylbenzylamine is categorized as a phenylmethylamine and naturally occurs in carrots, suggesting its potential use as a biomarker for the consumption of carrot-containing food products. It is noted for its susceptibility to spontaneous nitrosation, which increases under acidic conditions. The compound appears as a solid below -24°C and a liquid above -24°C, with a colorless to light yellow hue. For storage, it should be kept at 4°C, protected from light. This product is strictly for research use only.
  • Categorized as a phenylmethylamine
  • Naturally occurs in carrots
  • Potential use as a biomarker for carrot-containing food products
  • Susceptible to spontaneous nitrosation, enhanced under acidic conditions
  • Appears as a solid below -24°C and a liquid above -24°C
  • Colorless to light yellow hue
  • For research use only
Catalog No. 50-004-47851