Learn More
Medchemexpress LLC N-methylglycine-15N | 107-97-1 | MFCD00004279 | 99.9% | 90.09 g/mol | C3H7[15N]O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101037S1MG
Encompass_Preferred
15N-labeled sarcosine (N-methylglycine) is supplied as a high-purity stable isotope standard for tracer studies, quantitative mass spectrometry, and biochemical research into glycine transport and NMDA receptor modulation. It is intended for analytical workflows and small-scale experimental use.
  • 15N isotopic label for tracer and mass spectrometry applications.
  • High chemical and isotopic purity (>99.9%) for accurate quantitation.
  • Suitable as an internal standard for LC-MS/MS assays.
  • Compact package sizes for analytical-scale experiments.
  • Stable, white to off-white solid for convenient handling and storage.
Catalog No. 50-002-11977