Learn More
Medchemexpress LLC N-Methyliminodiacetic acid-d4 | 1219803-27-6 | 156.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW007872S5MG
Encompass_Preferred
2,2'-(Methylazanediyl)diacetic acid-d4 is a deuterium-labeled compound. Deuterium substitution in drug molecules is explored for its potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium-labeled compound
  • Used as a tracer for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-46451