Learn More
Medchemexpress LLC N-Methylisatoic anhydride | 10328-92-4 | 99.94% | 177.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1840710MM/1ML
Encompass_Preferred
N-Methylisatoic anhydride is a heteronucleoside anhydride derivative conjugated with biotin and a disulfide linker for specific 2'-OH acylation of RNA. Biotin-conjugated N-Methylisatoic anhydride can selectively capture RNA from a mixture of biological RNA and DNA. In vitro, N-Methylisatoic anhydride is a 2'-OH selective acylation agent of RNAs, widely used for resolving secondary RNA structures using the SHAPE (Selective 2'-Hydroxyl Acylation Analyzed by Primer Extension) technology.
  • Heteronucleoside anhydride derivative
  • Conjugated with biotin and a disulfide linker
  • Specific 2'-OH acylation of RNA
  • Selectively captures RNA from a mixture of biological RNA and DNA
  • 2'-OH selective acylation agent of RNAs
  • Used for resolving secondary RNA structures using SHAPE technology
Catalog No. 50-003-87835