Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylnicotinamide | 114-33-0 | MFCD00006384 | 99.6% | 136.15 g/mol | C7H8N2O | 5G
SDP

Supplier:  Medchemexpress LLC HY1241245G

Encompass_Preferred

N-Methylnicotinamide is an endogenous metabolite supplied as a high-purity solid for research use. It is provided as an analytical standard suitable for in vitro and in vivo studies, with recommended storage and formulation guidance for laboratory applications.

  • High purity: 99.6%
  • CAS number: 114-33-0
  • Molecular weight: 136.15 g/mol; formula: C7H8N2O
  • Appearance: white to off-white solid
  • Solubility: DMSO 200 mg/mL (may require sonication)
  • Recommended storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month)
  • Pack sizes: includes 5 G; larger quantities available upon request

Catalog No. 50-002-37771


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.