Learn More
Medchemexpress LLC N-Methylpiperazine-d4 | 343864-02-8 | 99.4% | C5H8D4N2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY78871S250MG
Encompass_Preferred
N-Methylpiperazine-d4 is the deuterium labeled N-Methylpiperazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33338