Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylprotoporphyrin IX | 79236-56-9 | MFCD08669600 | 99.0% | 576.68 | C35H36N4O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1294305MG

Encompass_Preferred

N-Methylprotoporphyrin IX is a porphyrin derivative and a potent inhibitor of the ferrochelatase enzyme used in research to study heme biosynthesis and enzyme inhibition. It is supplied as a high-purity solid suitable for biochemical assays and analytical applications.

  • Potent inhibitor of ferrochelatase enzyme.
  • High purity (99.0%).
  • Molecular formula C35H36N4O4 and molecular weight 576.68.
  • Solid, brown to black appearance.
  • Stable as powder at -20°C for extended storage.
  • Supplied in small research quantities (5 mg).
  • For research use only; not for human therapeutic or diagnostic use.

Catalog No. 50-002-62340


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.