Learn More
Medchemexpress LLC N-Methyltaurine 5g
SDP
Supplier:  Medchemexpress LLC HYW0983485G
Encompass_Preferred
N-Methyltaurine is a taurine-derived metabolite with a minimum effective concentration of 10 M for maintaining hamster sperm motility N-Methyltaurine is the dominant osmolyte of Vespermum virulentum[1][2]
Catalog No. 50-002-06431