Learn More
Medchemexpress LLC N,N'-1,3-phenylenebis[2,3-dihydroxy-benzamide] | 368449-04-1 | 98.6% | 50 MG

Supplier: Medchemexpress LLC HY12014550MG
MST-312 (Telomerase Inhibitor IX) is a small-molecule telomerase inhibitor derived from epigallocatechin gallate (EGCG). It is supplied as a white to off-white solid for laboratory research in oncology and telomerase studies.
- Telomerase inhibitor for cancer research.
- Chemically modified derivative of epigallocatechin gallate (EGCG).
- Molecular formula C20H16N2O6 and molecular weight 380.35 g·mol⁻1.
- Purity 98.6%.
- Supplied as a solid with common solubility in DMSO for solution preparation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.