Learn More
Strem, An Ascensus Company CAS# 172138-95-3. 1g. (+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide), min. 98% (S,S)-DACH-Pyridyl Trost Ligand. MFCD02684544
SDP
Supplier:  Strem, An Ascensus Company 0703411G
Encompass
CAS# 172138-95-3. 1g. (+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide), min. 98% (S,S)-DACH-Pyridyl Trost Ligand. MFCD02684544. Molecular Weight: 324.38. Molecular Formula: C18H20N4O2. Color/form: off-white pwdr. Strem# 07-0341. http://www.strem.com/catalog/v/07-0341/
Catalog No. 50-901-08983