Learn More
Medchemexpress LLC N,N-bis(2-chloroethyl)aniline | 553-27-5 | 99.4% | 25 G

Supplier: Medchemexpress LLC HY12171325G
Aniline mustard (N,N-bis(2-chloroethyl)aniline) is an anticancer alkylating agent reported to target complex I of the mitochondrial electron transport chain. It is supplied as an off-white to pale purple solid with high reported purity, documented solubility in DMSO, and manufacturer-provided formulation and storage guidance.
- High reported purity: 99.36% (manufacturer data)
- Solid form: off-white to pale purple, suitable for handling and formulation
- Solubility: highly soluble in DMSO; freshly opened DMSO recommended
- In vivo formulation protocols: examples provided for PEG300/Tween-80, SBE-β-CD, and corn oil vehicles
- Storage recommendations: store solid at 4°C protected from light; solutions long-term at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.