Learn More
Medchemexpress LLC N,N-Bis(2-hydroxyethyl)dodecanamide (Lauric diethanolamide) | 120-40-1 | 287.44 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0055135G
Encompass_Preferred
N,N-Bis(2-hydroxyethyl)dodecanamide (Lauric diethanolamide) is an orally consumable non-ionic surfactant and antistatic agent intended for research use only.
  • Orally consumable non-ionic surfactant
  • Antistatic agent
  • Solid appearance
  • Off-white to yellow color
  • Shipping at room temperature (continental US)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: ≥ 100 mg/mL (347.90 mM)
  • For research use only
Catalog No. 50-004-49028