Learn More
Strem, An Ascensus Company CAS# 2960-37-4. 1g. N,N-Bis(diphenylphosphino)amine, min. 98%. MFCD00233855
SDP
Supplier:  Strem, An Ascensus Company 1501101G
Encompass
CAS# 2960-37-4. 1g. N,N-Bis(diphenylphosphino)amine, min. 98%. MFCD00233855. Molecular Weight: 385.38. Molecular Formula: (C6H5)2PNHP(C6H5)2. Color/form: white xtl. Strem# 15-0110. http://www.strem.com/catalog/v/15-0110/
Catalog No. 50-901-09752