Learn More
AdipoGen N N-Bisphosphonomethylglycin
SDP
Supplier:  AdipoGen CDXN0054G050
Encompass_Preferred
Chemical. CAS 2439-99-8. Formula C4H11NO8P2. MW 263.08. Synthetic. A glycine-phosphonic acid compound, negatively charged at neutral pH, with immunomodulatory and anti-diabetic properties. Enhances anti-B9-23 TCR-dependent T cell activation by modulating murine class II MHC I-Ag7-mediated B9-23 presentation on APCs. Daily administration of glyphosine is shown to delay the onset of diabetes in NOD mice. Herbicide. A plant growth regulator. Acts as a competitive inhibitor of phoshoenolpyruvate carboxylase in plants.
Catalog No. 50-256-0423