Learn More
Medchemexpress LLC N,N'-Carbonyldiimidazole | 530-62-1 | 99.9% | C7H6N4O | 500 G
SDP
Supplier:  Medchemexpress LLC HY79880500G
Encompass_Preferred
N,N'-Carbonyldiimidazole (1,1'-Carbonyldiimidazole) is a highly reactive carboxylating reagent. It activates hydroxyl groups to form reactive carbonyl groups and functions as a coupling agent and peptide synthesis reagent. In vitro, it mediates the conjugation of ovalbumin with magnetite nanoparticles for vaccine delivery.
  • Highly reactive carboxylating reagent
  • Activates hydroxyl groups to form reactive carbonyl groups
  • Can be used as a coupling agent
  • Functions as a peptide synthesis reagent
  • Mediates conjugation of ovalbuminn with magnetite nanoparticles for vaccine delivery in vitro
Catalog No. 50-004-33188