Learn More
Medchemexpress LLC N,N'-Di-BOC-L-cystine | 10389-65-8 | ≥95.0% | 440.53 | 500 G

Supplier: Medchemexpress LLC HYW013734500G
N,N'-Di-BOC-L-cystine is a cysteine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: Powder: -20°C 3 years, 4°C 2 years; In solvent: -80°C 6 months, -20°C 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.