Learn More
Cayman Chemical N N-DIacetyl-L-cystIn 10g
SDP
Supplier:  Cayman Chemical 1759610 G
Encompass_Preferred
A disulfide dimer of N-acetylcysteine with immunomodulatory and anti-atherosclerotic properties; modifies contact sensitivity/delayed hypersensitivity reactions in mice and improves endothelial function in Watanabe heritable hyperlipidemic rabbits
Catalog No. 50-296-6846