Learn More
Medchemexpress LLC N,N'-dicyclohexylcarbodiimide (DCC) | 538-75-0 | MFCD00011659 | 99.4% | 206.33 g/mol | C13H22N2 | 25g
SDP
Supplier:  Medchemexpress LLC HYY109225G
Encompass_Preferred
N N-Dicyclohexylcarbodiimide(DCC) is a dehydrating and condensing agent N N-Dicyclohexylcarbodiimide(DCC) enhances adenylate cyclase activity N N-Dicyclohexylcarbodiimide(DCC) inhibits amine accumulation N N-Dicyclohexylcarbodiimide(DCC) is used to prepare 3-phenylcoumarin derivatives sulfated derivatives of purslane polysaccharides[1][2][3][4][5][6]
Catalog No. 50-002-67489