Learn More
Medchemexpress LLC N,N'-Diferuloylputrescine | 42369-86-8 | 440.49 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN122671MG
Encompass_Preferred
N,N′-Diferuloylputrescine is a pigmentation inhibitor that causes a 57% reduction in pigmentation. It works by significantly reducing the protein level of MITF. Additionally, N,N′-Diferuloylputrescine exhibits strong antioxidant activities by acting as a radical scavenger against reactive oxygen species.
  • Inhibitor of pigmentation with 57% reduction
  • Significantly reduces the protein level of MITF
  • Possesses strong antioxidant activities as a radical scavenger against reactive oxygen species
Catalog No. 50-004-14597